Alfa Aesar™ 2-(2-Methoxyphenoxy)ethylamine, 97%

Manufacturer: Alfa Aesar™ H5025103

Quantity 1g

 View more versions of this product

Catalog No. AAH5025103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1836-62-0
    Molecular Formula C9H13NO2
    Formula Weight 167.21
    MDL Number MFCD00235185
    UN Number UN2735
    Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine

    Specifications

    Formula Weight 167.21
    Molecular Formula C9H13NO2
    MDL Number MFCD00235185
    Boiling Point 98°C (0.4mmHg)
    CAS 1836-62-0
    UN Number UN2735
    Quantity 1g
    Odor Amine-like
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine
    Assay 97%

    Description

    Applications
    2-(2-Methoxyphenoxy)ethylamine is used for its reaction with 4-(2,3-epoxypropoxy) carbazole for preparation of Carvedilol (I).

    Solubility
    Difficult to mix.

    Notes
    Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit