Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Methoxyphenyl)benzimidazole, 95%

Click to view available options
Quantity:
250mg
1g
5g
6528-85-4
C14H12N2O
224.263
MFCD00447053
JNFBDHNAZPBWTF-UHFFFAOYSA-N
2-2-methoxyphenyl-1h-benzimidazole,1h-benzimidazole, 2-2-methoxyphenyl,2-2-methoxyphenyl-1h-benzo d imidazole,2-2-methoxyphenyl-1h-1,3-benzodiazole,2-2-methoxy-phenyl-1h-benzoimidazole,1h-benzo d imidazole,2-2-methoxyphenyl
759420
2-(2-methoxyphenyl)-1H-benzimidazole
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
6528-85-4
C14H12N2O
224.263
MFCD00447053
JNFBDHNAZPBWTF-UHFFFAOYSA-N
2-2-methoxyphenyl-1h-benzimidazole,1h-benzimidazole, 2-2-methoxyphenyl,2-2-methoxyphenyl-1h-benzo d imidazole,2-2-methoxyphenyl-1h-1,3-benzodiazole,2-2-methoxy-phenyl-1h-benzoimidazole,1h-benzo d imidazole,2-2-methoxyphenyl
759420
2-(2-methoxyphenyl)-1H-benzimidazole
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
Quantity 1g
Formula Weight 224.26
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-(2-Methoxyphenyl)benzimidazole