Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine, 98%

Click to view available options
Quantity:
1g
5g
25g
152460-09-8
C16H13N5O2
307.313
MFCD02179269
OJITWRFPRCHSMX-UHFFFAOYSA-N
n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine,n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine,chembl20426,n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine,n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine,2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine,2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine,2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl,n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid,n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
10686149
N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3
152460-09-8
C16H13N5O2
307.313
MFCD02179269
OJITWRFPRCHSMX-UHFFFAOYSA-N
n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine,n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine,chembl20426,n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine,n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine,2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine,2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine,2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl,n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid,n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
10686149
N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3
Quantity 5g
Melting Point 188°C to 193°C
Color Yellow
Beilstein 7544032
Formula Weight 307.31
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning