Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Methyl-6-nitrophenyl)acetic acid, 95%

Click to view available options
Quantity:
250mg
1g
23876-18-8
C9H9NO4
195.174
MFCD00066313
WFKNETIJXXWRHO-UHFFFAOYSA-N
2-2-methyl-6-nitrophenyl acetic acid,2-methyl-6-nitrophenylacetic acid,2-methyl-6-nitrophenyl acetic acid,2-methyl-6-nitro-phenylacetic acid,2-6-methyl-2-nitrophenyl acetic acid
675115
CC1=C(C(=CC=C1)[N+](=O)[O-])CC(=O)O
23876-18-8
C9H9NO4
195.174
MFCD00066313
WFKNETIJXXWRHO-UHFFFAOYSA-N
2-2-methyl-6-nitrophenyl acetic acid,2-methyl-6-nitrophenylacetic acid,2-methyl-6-nitrophenyl acetic acid,2-methyl-6-nitro-phenylacetic acid,2-6-methyl-2-nitrophenyl acetic acid
675115
CC1=C(C(=CC=C1)[N+](=O)[O-])CC(=O)O
Quantity 250mg
Beilstein 2617187
Formula Weight 195.17
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-(2-Methyl-6-nitrophenyl)acetic acid