Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Naphthoyl)-1-(o-toluoyl)hydrazine, 98%

Click to view available options
Quantity:
1g
5g
83803-94-5
C19H16N2O2
304.349
MFCD00209665
PLZLECZNFLCRPC-UHFFFAOYSA-N
2-2-naphthoyl-1-o-toluoyl hydrazine,n'-2-methylbenzoyl-2-naphthohydrazide,2'-2-methylbenzoyl-2-naphthohydrazide,n'-2-methylbenzoyl naphthalene-2-carbohydrazide,cbmicro_017697,1-2-methylbenzoyl-2-2-naphthoyl hydrazine,n'-2-methylbenzoyl-2-naphthalenecarbohydrazide,2-methyl-n'-naphthalene-2-carbonyl benzohydrazide,2-methylphenyl-n-2-naphthylcarbonylamino carboxamide
96760
N'-(2-methylbenzoyl)naphthalene-2-carbohydrazide
CC1=CC=CC=C1C(=O)NNC(=O)C2=CC3=CC=CC=C3C=C2
83803-94-5
C19H16N2O2
304.349
MFCD00209665
PLZLECZNFLCRPC-UHFFFAOYSA-N
2-2-naphthoyl-1-o-toluoyl hydrazine,n'-2-methylbenzoyl-2-naphthohydrazide,2'-2-methylbenzoyl-2-naphthohydrazide,n'-2-methylbenzoyl naphthalene-2-carbohydrazide,cbmicro_017697,1-2-methylbenzoyl-2-2-naphthoyl hydrazine,n'-2-methylbenzoyl-2-naphthalenecarbohydrazide,2-methyl-n'-naphthalene-2-carbonyl benzohydrazide,2-methylphenyl-n-2-naphthylcarbonylamino carboxamide
96760
N'-(2-methylbenzoyl)naphthalene-2-carbohydrazide
CC1=CC=CC=C1C(=O)NNC(=O)C2=CC3=CC=CC=C3C=C2
Quantity 1g
Formula Weight 304.33
Percent Purity 98%
Chemical Name or Material 2-(2-Naphthoyl)-1-(o-toluoyl)hydrazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning