Alfa Aesar™ 2-(2-Pyrrolidinyl)pyridine, 96%

Manufacturer: Alfa Aesar™ H50176MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50176MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77790-61-5
    Molecular Formula C9H12N2
    Formula Weight 148.21
    MDL Number MFCD01691538
    UN Number UN2810
    Synonym alpha-Nornicotine; 2-(2-Pyridyl)pyrrolidine
    Chemical Name or Material 2-(2-Pyrrolidinyl)pyridine

    Specifications

    Formula Weight 148.21
    Molecular Formula C9H12N2
    MDL Number MFCD01691538
    CAS 77790-61-5
    UN Number UN2810
    Synonym alpha-Nornicotine; 2-(2-Pyridyl)pyrrolidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    RTECS Number UT7650000
    Quantity 250mg
    Chemical Name or Material 2-(2-Pyrrolidinyl)pyridine
    Assay 96%

    Description

    Applications
    2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

    Solubility
    Soluble in water.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit