Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2'-Thenoin, 97%

Click to view available options
Quantity:
1g
5g
25g
27761-02-0
C10H8O2S2
224.292
MFCD00022505
OGWZIOZTPNLTCR-UHFFFAOYSA-N
2,2'-thenoin,2-hydroxy-1,2-di-2-thienylethanone,2-hydroxy-1,2-di 2-thienyl ethan-1-one,2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one,2-hydroxy-1,2-di thiophen-2-yl ethanone,2-hydroxy-1,2-bis thiophen-2-yl ethanone,maybridge1_002119,1,2-dithien-2-ylhydroxyethanone,2-hydroxy-1,2-dithien-2-ylethanone,alpha-hydroxy-2-thenyl 2-thienyl ketone
2734800
2-hydroxy-1,2-dithiophen-2-ylethanone
C1=CSC(=C1)C(C(=O)C2=CC=CS2)O
27761-02-0
C10H8O2S2
224.292
MFCD00022505
OGWZIOZTPNLTCR-UHFFFAOYSA-N
2,2'-thenoin,2-hydroxy-1,2-di-2-thienylethanone,2-hydroxy-1,2-di 2-thienyl ethan-1-one,2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one,2-hydroxy-1,2-di thiophen-2-yl ethanone,2-hydroxy-1,2-bis thiophen-2-yl ethanone,maybridge1_002119,1,2-dithien-2-ylhydroxyethanone,2-hydroxy-1,2-dithien-2-ylethanone,alpha-hydroxy-2-thenyl 2-thienyl ketone
2734800
2-hydroxy-1,2-dithiophen-2-ylethanone
C1=CSC(=C1)C(C(=O)C2=CC=CS2)O
Melting Point 104°C to 107°C
Quantity 25g
Formula Weight 224.29
Percent Purity 97%
Chemical Name or Material 2,2'-Thenoin