Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2′-Thenoin 97.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
27761-02-0
C10H8O2S2
224.292
MFCD00022505
OGWZIOZTPNLTCR-UHFFFAOYSA-N
2,2'-thenoin,2-hydroxy-1,2-di-2-thienylethanone,2-hydroxy-1,2-di 2-thienyl ethan-1-one,2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one,2-hydroxy-1,2-di thiophen-2-yl ethanone,2-hydroxy-1,2-bis thiophen-2-yl ethanone,maybridge1_002119,1,2-dithien-2-ylhydroxyethanone,2-hydroxy-1,2-dithien-2-ylethanone,alpha-hydroxy-2-thenyl 2-thienyl ketone
2734800
2-hydroxy-1,2-dithiophen-2-ylethanone
C1=CSC(=C1)C(C(=O)C2=CC=CS2)O
27761-02-0
C10H8O2S2
224.292
MFCD00022505
OGWZIOZTPNLTCR-UHFFFAOYSA-N
2,2'-thenoin,2-hydroxy-1,2-di-2-thienylethanone,2-hydroxy-1,2-di 2-thienyl ethan-1-one,2-hydroxy-1,2-bis thiophen-2-yl ethan-1-one,2-hydroxy-1,2-di thiophen-2-yl ethanone,2-hydroxy-1,2-bis thiophen-2-yl ethanone,maybridge1_002119,1,2-dithien-2-ylhydroxyethanone,2-hydroxy-1,2-dithien-2-ylethanone,alpha-hydroxy-2-thenyl 2-thienyl ketone
2734800
2-hydroxy-1,2-dithiophen-2-ylethanone
C1=CSC(=C1)C(C(=O)C2=CC=CS2)O
Color Yellow
Quantity 1g
Melting Point 109°C
Formula Weight 224.29
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (HPLC)
Chemical Name or Material 2,2′-Thenoin