Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Thienyl)ethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5402-55-1
C6H8OS
128.19
MFCD00005462
VMJOFTHFJMLIKL-UHFFFAOYSA-N
2-thiopheneethanol,2-2-thienyl ethanol,thiophene-2-ethanol,2-thiophen-2-yl ethanol,2-2-hydroxyethyl thiophene,2-thiophen-2-yl ethan-1-ol,ethanol, 2-2-thienyl,2-thiophenethanol,2-2-thienyl-ethanol,2-thienylethanol
79400
2-thiophen-2-ylethanol
C1=CSC(=C1)CCO
5402-55-1
C6H8OS
128.19
MFCD00005462
VMJOFTHFJMLIKL-UHFFFAOYSA-N
2-thiopheneethanol,2-2-thienyl ethanol,thiophene-2-ethanol,2-thiophen-2-yl ethanol,2-2-hydroxyethyl thiophene,2-thiophen-2-yl ethan-1-ol,ethanol, 2-2-thienyl,2-thiophenethanol,2-2-thienyl-ethanol,2-thienylethanol
79400
2-thiophen-2-ylethanol
C1=CSC(=C1)CCO
Refractive Index 1.5490 to 1.552
Density 1.1530g/mL
Boiling Point 108°C (13.0mmHg)
Flash Point 101°C
Specific Gravity 1.153
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown or Gray-Green to Yellow
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 17, V,3, 379
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in mek,alcohol,organic halogen solvents
Formula Weight 128.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-(2-Thienyl)ethanol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-452-

Recommended Storage : this product discolors on storage