Alfa Aesar™ 2,2'-Thiobisacetamide, 97%

Manufacturer: Alfa Aesar™ B25254-03

Quantity 1g

 View more versions of this product

Catalog No. AAB2525403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14618-65-6
    Molecular Formula S(CH2CONH2)2
    Formula Weight 148.18g/mol
    MDL Number MFCD00014806
    Synonym 2,2'-Thiodiacetamide
    Chemical Name or Material 2,2'-Thiobisacetamide

    Specifications

    Melting Point 172° to 176°C
    Quantity 1g
    CAS 14618-65-6
    Molecular Formula S(CH2CONH2)2
    Formula Weight 148.18g/mol
    MDL Number MFCD00014806
    Synonym 2,2'-Thiodiacetamide
    Chemical Name or Material 2,2'-Thiobisacetamide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit