Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2′-Thiodiglycolic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
123-93-3
C4H6O4S
150.148
MFCD00004359
UVZICZIVKIMRNE-UHFFFAOYSA-N
thiodiglycolic acid,2,2'-thiodiacetic acid,thiodiacetic acid,mercaptodiacetic acid,tdga,dicarboxymethyl sulfide,thiodi acetic acid,carboxymethylthio acetic acid,2,2'-thiodiethanoic acid,thiodiglycollic acid
31277
CHEBI:35012
2-(carboxymethylsulfanyl)acetic acid
C(C(=O)O)SCC(=O)O
123-93-3
C4H6O4S
150.148
MFCD00004359
UVZICZIVKIMRNE-UHFFFAOYSA-N
thiodiglycolic acid,2,2'-thiodiacetic acid,thiodiacetic acid,mercaptodiacetic acid,tdga,dicarboxymethyl sulfide,thiodi acetic acid,carboxymethylthio acetic acid,2,2'-thiodiethanoic acid,thiodiglycollic acid
31277
CHEBI:35012
2-(carboxymethylsulfanyl)acetic acid
C(C(=O)O)SCC(=O)O
Quantity 500g
Color Undesignated
Melting Point 129°C
UN Number 3261
Formula Weight 150.15
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,2′-Thiodiglycolic Acid

RTECSNumber : AJ6475000

TSCA : Yes