Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
774-18-5
C9H10F3N
189.181
MFCD00128232
LIERORLYMWHXDL-UHFFFAOYSA-N
2-2-trifluoromethyl phenyl ethanamine,o-trifluoromethyl phenethylamine,2-2-trifluoromethyl phenyl ethan-1-amine,2-o-trifluoromethylphenyl ethylamine,2-2-trifluoromethylphenyl ethylamine,2-trifluoromethyl phenethylamine,2-2-trifluoromethyl-phenyl-ethylamine,2-2-trifluoromethyl phenyl ethyl amine,2-trifluoromethylphenylethanamine,benzeneethanamine,2-trifluoromethyl
69883
2-[2-(trifluoromethyl)phenyl]ethanamine
C1=CC=C(C(=C1)CCN)C(F)(F)F
774-18-5
C9H10F3N
189.181
MFCD00128232
LIERORLYMWHXDL-UHFFFAOYSA-N
2-2-trifluoromethyl phenyl ethanamine,o-trifluoromethyl phenethylamine,2-2-trifluoromethyl phenyl ethan-1-amine,2-o-trifluoromethylphenyl ethylamine,2-2-trifluoromethylphenyl ethylamine,2-trifluoromethyl phenethylamine,2-2-trifluoromethyl-phenyl-ethylamine,2-2-trifluoromethyl phenyl ethyl amine,2-trifluoromethylphenylethanamine,benzeneethanamine,2-trifluoromethyl
69883
2-[2-(trifluoromethyl)phenyl]ethanamine
C1=CC=C(C(=C1)CCN)C(F)(F)F
Quantity 25g
Color Yellow
UN Number UN2735
Beilstein 7202211
Formula Weight 189.18
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-[2-(Trifluoromethyl)phenyl]ethylamine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.