Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,3',4'-Tetramethoxybenzophenone, 98%, Acros Organics

Click to view available options
Quantity:
5g
50625-53-1
C17H18O5
302.326
ZZMLFJJROAGMSL-UHFFFAOYSA-N
2,3,3',4'-tetramethoxybenzophenone,2,3-dimethoxyphenyl 3,4-dimethoxyphenyl methanone,di2,3-dimethoxyphenyl ketone,2,3,3'4'-tetramethoxybenzophenone,2,3-dimethoxyphenyl-3,4-dimethoxyphenyl methanone,methanone, 2,3-dimethoxyphenyl 3,4-dimethoxyphenyl,2,3-dimethoxyphenyl 3,4-dimethoxyphenyl methanone 6
2724108
(2,3-dimethoxyphenyl)-(3,4-dimethoxyphenyl)methanone
COC1=C(C=C(C=C1)C(=O)C2=C(C(=CC=C2)OC)OC)OC
50625-53-1
C17H18O5
302.326
ZZMLFJJROAGMSL-UHFFFAOYSA-N
2,3,3',4'-tetramethoxybenzophenone,2,3-dimethoxyphenyl 3,4-dimethoxyphenyl methanone,di2,3-dimethoxyphenyl ketone,2,3,3'4'-tetramethoxybenzophenone,2,3-dimethoxyphenyl-3,4-dimethoxyphenyl methanone,methanone, 2,3-dimethoxyphenyl 3,4-dimethoxyphenyl,2,3-dimethoxyphenyl 3,4-dimethoxyphenyl methanone 6
2724108
(2,3-dimethoxyphenyl)-(3,4-dimethoxyphenyl)methanone
COC1=C(C=C(C=C1)C(=O)C2=C(C(=CC=C2)OC)OC)OC
Quantity 5g
Melting Point 118°C to 120°C
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula {(CH3O)2C6H3}2CO
Formula Weight 302.32
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,3,3',4'-Tetramethoxybenzophenone, 98%
Show More Show Less