Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,3-Trimethyl-1-butene 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
594-56-9
C7H14
98.19
MFCD00008850
AUYRUAVCWOAHQN-UHFFFAOYSA-N
2,3,3-trimethyl-1-butene,1-butene, 2,3,3-trimethyl,triptene,acmc-209mdj,2,3-trimethyl-1-butene,1-butene,3,3-trimethyl,1-butene,2,3,3-trimethyl,ch3 3cc ch3 =ch2
11669
2,3,3-trimethylbut-1-ene
CC(=C)C(C)(C)C
594-56-9
C7H14
98.19
MFCD00008850
AUYRUAVCWOAHQN-UHFFFAOYSA-N
2,3,3-trimethyl-1-butene,1-butene, 2,3,3-trimethyl,triptene,acmc-209mdj,2,3-trimethyl-1-butene,1-butene,3,3-trimethyl,1-butene,2,3,3-trimethyl,ch3 3cc ch3 =ch2
11669
2,3,3-trimethylbut-1-ene
CC(=C)C(C)(C)C
Refractive Index 1.4015 to 1.4035
Boiling Point 78°C to 80°C
Flash Point -17°C
Specific Gravity 0.7
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Density 0.7000g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula (CH3)3CC(CH3)=CH2
Beilstein 01, 221
Formula Weight 98.19
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material 2, 3, 3-Trimethyl-1-butene, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-845-1

TSCA : TSCA