Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,3-Trimethylindolenine, 98%

Click to view available options
Quantity:
5g
25g
1640-39-7
C11H13N
159.232
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine,2,3,3-trimethyl-3h-indole,3h-indole, 2,3,3-trimethyl,ccris 6607,2,3,3-trimethyl-indolenin,2,3,3-trimethyl-3-hydroindole,pubchem7445,rarechem ah bs 0130,2,3-trimethylindolenine,2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C
1640-39-7
C11H13N
159.232
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine,2,3,3-trimethyl-3h-indole,3h-indole, 2,3,3-trimethyl,ccris 6607,2,3,3-trimethyl-indolenin,2,3,3-trimethyl-3-hydroindole,pubchem7445,rarechem ah bs 0130,2,3-trimethylindolenine,2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C
Applications
It is used in synthesis of cyanine dyes.

Solubility
Soluble in chloroform, toluene or dichlorobenzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Melting Point 6°C to 8°C
Boiling Point 228°C to 229°C
Quantity 5g
Flash Point 93°C (199°F)
Beilstein 119740
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material 2,3,3-Trimethylindolenine

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA