Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,3-Trimethylindolenine 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
Packaging:
Glass bottle
1640-39-7
C11H13N
159.23
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine,2,3,3-trimethyl-3h-indole,3h-indole, 2,3,3-trimethyl,ccris 6607,2,3,3-trimethyl-indolenin,2,3,3-trimethyl-3-hydroindole,pubchem7445,rarechem ah bs 0130,2,3-trimethylindolenine,2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C
1640-39-7
C11H13N
159.23
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine,2,3,3-trimethyl-3h-indole,3h-indole, 2,3,3-trimethyl,ccris 6607,2,3,3-trimethyl-indolenin,2,3,3-trimethyl-3-hydroindole,pubchem7445,rarechem ah bs 0130,2,3-trimethylindolenine,2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C
Density 0.9900g/mL
Flash Point 93°C
Boiling Point 228°C to 229°C (744.0mmHg)
Refractive Index 1.545
Specific Gravity 0.99
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Undesignated
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 20, 324
Formula Weight 159.23
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 3, 3-Trimethylindolenine, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 216-685-6

TSCA : TSCA