Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4',5',6'-Pentafluoroacetophenone, 97%, Acros Organics

Click to view available options
Quantity:
2.5g
10g
652-29-9
C8H3F5O
210.103
MFCD00000296
FBGHCYZBCMDEOX-UHFFFAOYSA-N
2',3',4',5',6'-pentafluoroacetophenone,1-perfluorophenyl ethanone,pentafluoroacetophenone,1-pentafluorophenyl ethan-1-one,ethanone, 1-pentafluorophenyl,2,3,4,5,6-pentafluoroacetophenone,1-pentafluorphenyl ethanon,1-acetyl-2,3,4,5,6-pentafluorobenzene,acmc-209npz,1-pentafluorophenylethanone
69546
1-(2,3,4,5,6-pentafluorophenyl)ethanone
CC(=O)C1=C(C(=C(C(=C1F)F)F)F)F
652-29-9
C8H3F5O
210.103
MFCD00000296
FBGHCYZBCMDEOX-UHFFFAOYSA-N
2',3',4',5',6'-pentafluoroacetophenone,1-perfluorophenyl ethanone,pentafluoroacetophenone,1-pentafluorophenyl ethan-1-one,ethanone, 1-pentafluorophenyl,2,3,4,5,6-pentafluoroacetophenone,1-pentafluorphenyl ethanon,1-acetyl-2,3,4,5,6-pentafluorobenzene,acmc-209npz,1-pentafluorophenylethanone
69546
1-(2,3,4,5,6-pentafluorophenyl)ethanone
CC(=O)C1=C(C(=C(C(=C1F)F)F)F)F
Color Colorless to Yellow
Quantity 2.5g
Infrared Spectrum Authentic
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Packaging Glass bottle
Density 1.47g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6F5COCH3
Formula Weight 210.1
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2',3',4',5',6'-Pentafluoroacetophenone, 97%
Show More Show Less

EINECSNumber : 211-487-6

TSCA : TSCA