Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluoroaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
250g
Packaging:
Glass bottle
771-60-8
C6H2F5N
183.081
MFCD00007643
NOXLGCOSAFGMDV-UHFFFAOYSA-N
pentafluoroaniline,pentafluorophenylamine,aminopentafluorobenzene,benzenamine, 2,3,4,5,6-pentafluoro,2,3,4,5,6-pentafluorobenzenamine,aniline, 2,3,4,5,6-pentafluoro,unii-ms06il3y2q,ms06il3y2q,2,3,4,5,6-pentafluorophenylamine,pubchem1546
13040
2,3,4,5,6-pentafluoroaniline
C1(=C(C(=C(C(=C1F)F)F)F)F)N
771-60-8
C6H2F5N
183.081
MFCD00007643
NOXLGCOSAFGMDV-UHFFFAOYSA-N
pentafluoroaniline,pentafluorophenylamine,aminopentafluorobenzene,benzenamine, 2,3,4,5,6-pentafluoro,2,3,4,5,6-pentafluorobenzenamine,aniline, 2,3,4,5,6-pentafluoro,unii-ms06il3y2q,ms06il3y2q,2,3,4,5,6-pentafluorophenylamine,pubchem1546
13040
2,3,4,5,6-pentafluoroaniline
C1(=C(C(=C(C(=C1F)F)F)F)F)N
Melting Point 33°C to 38°C
Color Salmon to White
Density 73g/mL
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6F5NH2
Formula Weight 183.08
Physical Form Crystalline Low Melting Solid
Percent Purity ≥96.0%
Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning