Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4,5,6-Pentafluoroanisole, 98%

Click to view available options
Quantity:
5g
25g
100g
389-40-2
C7H3F5O
198.092
MFCD00000289
ZRQUIRABLIQJRI-UHFFFAOYSA-N
2,3,4,5,6-pentafluoroanisole,pentafluoroanisole,benzene, pentafluoromethoxy,pentafluorophenyl methyl ether,pentafluoromethoxybenzene,methyl pentafluorophenyl ether,benzene, 1,2,3,4,5-pentafluoro-6-methoxy,anisole, 2,3,4,5,6-pentafluoro,1,2,3,4,6-pentafluoro-5-methoxybenzene,methoxypentafluorobenzene
67849
1,2,3,4,5-pentafluoro-6-methoxybenzene
COC1=C(C(=C(C(=C1F)F)F)F)F
389-40-2
C7H3F5O
198.092
MFCD00000289
ZRQUIRABLIQJRI-UHFFFAOYSA-N
2,3,4,5,6-pentafluoroanisole,pentafluoroanisole,benzene, pentafluoromethoxy,pentafluorophenyl methyl ether,pentafluoromethoxybenzene,methyl pentafluorophenyl ether,benzene, 1,2,3,4,5-pentafluoro-6-methoxy,anisole, 2,3,4,5,6-pentafluoro,1,2,3,4,6-pentafluoro-5-methoxybenzene,methoxypentafluorobenzene
67849
1,2,3,4,5-pentafluoro-6-methoxybenzene
COC1=C(C(=C(C(=C1F)F)F)F)F
Quantity 5g
Boiling Point 136°C to 138°C
Melting Point -38°C
Flash Point 32°C (89°F)
UN Number UN1993
Beilstein 2052664
Formula Weight 198.09
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluoroanisole

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA