Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ B2292214

Quantity 25g

 View more versions of this product

Catalog No. AAB2292214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1582-24-7
    Molecular Formula C6H2BF5O2
    Formula Weight 211.88
    MDL Number MFCD01074663
    Beilstein 3033960
    Synonym 2,3,4,5,6-Pentafluorophenylboronic acid
    Chemical Name or Material 2,3,4,5,6-Pentafluorobenzeneboronic acid

    Specifications

    Formula Weight 211.88
    Melting Point 250° to 255°C
    Molecular Formula C6H2BF5O2
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD01074663
    CAS 1582-24-7
    Beilstein 3033960
    Synonym 2,3,4,5,6-Pentafluorophenylboronic acid
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2,3,4,5,6-Pentafluorobenzeneboronic acid
    Assay 97%

    Description

    Applications
    2,3,4,5,6-Pentafluorobenzeneboronic acid is utilized in the palladium-catalyzed Suzuki reaction to synthesize several cross-coupled aromatic compounds, for example bis(pentafluorophenyl)-substituted thiophene and selenophene derivatives. It serves as an intermediate and raw material in pharmaceutical and chemical industry. It is also used in dyes and pigments. It can be used to prepare cyclic boronates of bifunctional compounds.

    Solubility
    Miscible with alcohols, acetonitrile and dichloromethane.

    Notes
    Moisture Sensitive. Store in cool and dry conditions in well sealed containers. Keep container tightly sealed.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit