Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzeneboronic acid, 97%

Click to view available options
Quantity:
5g
25g
1582-24-7
C6H2BF5O2
211.882
MFCD01074663
VASOMTXTRMYSKD-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzeneboronic acid,pentafluorophenylboronic acid,perfluorophenylboronic acid,pentafluorobenzeneboronic acid,boronic acid, pentafluorophenyl,pentafluorophenyl boronic acid,2,3,4,5,6-pentafluorophenyl boronic acid,boronic acid, b-2,3,4,5,6-pentafluorophenyl,pubchem22760,rarechem ah pb 0049
2775922
(2,3,4,5,6-pentafluorophenyl)boronic acid
B(C1=C(C(=C(C(=C1F)F)F)F)F)(O)O
1582-24-7
C6H2BF5O2
211.882
MFCD01074663
VASOMTXTRMYSKD-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzeneboronic acid,pentafluorophenylboronic acid,perfluorophenylboronic acid,pentafluorobenzeneboronic acid,boronic acid, pentafluorophenyl,pentafluorophenyl boronic acid,2,3,4,5,6-pentafluorophenyl boronic acid,boronic acid, b-2,3,4,5,6-pentafluorophenyl,pubchem22760,rarechem ah pb 0049
2775922
(2,3,4,5,6-pentafluorophenyl)boronic acid
B(C1=C(C(=C(C(=C1F)F)F)F)F)(O)O
Applications
2,3,4,5,6-Pentafluorobenzeneboronic acid is utilized in the palladium-catalyzed Suzuki reaction to synthesize several cross-coupled aromatic compounds, for example bis(pentafluorophenyl)-substituted thiophene and selenophene derivatives. It serves as an intermediate and raw material in pharmaceutical and chemical industry. It is also used in dyes and pigments. It can be used to prepare cyclic boronates of bifunctional compounds.

Solubility
Miscible with alcohols, acetonitrile and dichloromethane.

Notes
Moisture Sensitive. Store in cool and dry conditions in well sealed containers. Keep container tightly sealed.
Melting Point 250°C to 255°C
Quantity 5g
Beilstein 3033960
Formula Weight 211.88
Percent Purity 97%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning