Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzyl alcohol, 98%

Manufacturer: Alfa Aesar™ A1211914

Quantity 25g

 View more versions of this product

Catalog No. AAA1211914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 440-60-8
    Molecular Formula C7H3F5O
    Formula Weight 198.09
    MDL Number MFCD00004602
    Beilstein 2052669
    Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol

    Specifications

    Formula Weight 198.09
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 32° to 36°C
    Molecular Formula C7H3F5O
    Beilstein 2052669
    RTECS Number DP0695000
    Boiling Point 182°C
    Quantity 25g
    Flash Point 87°C (188°F)
    MDL Number MFCD00004602
    CAS 440-60-8
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol
    Assay 98%

    GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit