Alfa Aesar™ 2,3,4,5,6-Pentafluorobenzyl bromide, 98+%

Manufacturer: Alfa Aesar™ A1162314

Quantity 25g

 View more versions of this product

Catalog No. AAA1162314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1765-40-8
    Molecular Formula C7H2BrF5
    Formula Weight 261
    MDL Number MFCD00000299
    UN Number UN3265
    Beilstein 647808
    Synonym alpha-Bromo-2,3,4,5,6-pentafluorotoluene
    Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl bromide

    Specifications

    Formula Weight 261
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00000299
    Health Hazard 1 Danger
    Boiling Point 174° to 175°C
    Molecular Formula C7H2BrF5
    Flash Point 83°C (181°F)
    CAS 1765-40-8
    UN Number UN3265
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Melting Point 18° to 20°C
    Quantity 25g
    Beilstein 647808
    Synonym alpha-Bromo-2,3,4,5,6-pentafluorotoluene
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl bromide
    Assay ≥98%

    Description

    Applications
    2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

    Solubility
    Hydrolyzes in water.

    Notes
    Store in a cool, dry, well-ventilated area. Incompatible with bases and oxidizing agents. Store at 4°C. Protected from moisture.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit