Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorocinnamic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
719-60-8
C9H3F5O2
238.113
MFCD00004371
IUUKDBLGVZISGW-OWOJBTEDSA-N
2,3,4,5,6-pentafluorocinnamic acid,2e-3-pentafluorophenyl prop-2-enoic acid,3-perfluorophenyl acrylic acid,2e-3-2,3,4,5,6-pentafluorophenyl prop-2-enoic acid,3-pentafluorophenyl acrylic acid,2,3,4,5,6-pentafluorocinnamic aci,e-3-2,3,4,5,6-pentafluorophenyl prop-2-enoic acid,2e-3-2,3,4,5,6-pentafluorophenyl-2-propenoic acid #
1796494
(E)-3-(2,3,4,5,6-pentafluorophenyl)prop-2-enoic acid
C(=CC(=O)O)C1=C(C(=C(C(=C1F)F)F)F)F
719-60-8
C9H3F5O2
238.113
MFCD00004371
IUUKDBLGVZISGW-OWOJBTEDSA-N
2,3,4,5,6-pentafluorocinnamic acid,2e-3-pentafluorophenyl prop-2-enoic acid,3-perfluorophenyl acrylic acid,2e-3-2,3,4,5,6-pentafluorophenyl prop-2-enoic acid,3-pentafluorophenyl acrylic acid,2,3,4,5,6-pentafluorocinnamic aci,e-3-2,3,4,5,6-pentafluorophenyl prop-2-enoic acid,2e-3-2,3,4,5,6-pentafluorophenyl-2-propenoic acid #
1796494
(E)-3-(2,3,4,5,6-pentafluorophenyl)prop-2-enoic acid
C(=CC(=O)O)C1=C(C(=C(C(=C1F)F)F)F)F
Quantity 1g
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 150°C to 154°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6F5CH=CHCO2H
Formula Weight 238.11
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,3,4,5,6-Pentafluorocinnamic acid, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.