Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4,5,6-Pentafluorophenylacetic acid, 98+%

Click to view available options
Quantity:
1g
5g
653-21-4
C8H3F5O2
226.102
MFCD00004316
LGCODSNZJOVMHV-UHFFFAOYSA-N
pentafluorophenylacetic acid,2,3,4,5,6-pentafluorophenylacetic acid,pentafluorophenyl acetic acid,2-perfluorophenyl acetic acid,2-2,3,4,5,6-pentafluorophenyl acetic acid,2,3,4,5,6-pentafluorphenylacetic acid,benzeneacetic acid, 2,3,4,5,6-pentafluoro,acmc-20aoj2,pentafluorophenylaceticacid,2-pentafluorophenyl acetic acid
69553
2-(2,3,4,5,6-pentafluorophenyl)acetic acid
C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O
653-21-4
C8H3F5O2
226.102
MFCD00004316
LGCODSNZJOVMHV-UHFFFAOYSA-N
pentafluorophenylacetic acid,2,3,4,5,6-pentafluorophenylacetic acid,pentafluorophenyl acetic acid,2-perfluorophenyl acetic acid,2-2,3,4,5,6-pentafluorophenyl acetic acid,2,3,4,5,6-pentafluorphenylacetic acid,benzeneacetic acid, 2,3,4,5,6-pentafluoro,acmc-20aoj2,pentafluorophenylaceticacid,2-pentafluorophenyl acetic acid
69553
2-(2,3,4,5,6-pentafluorophenyl)acetic acid
C(C1=C(C(=C(C(=C1F)F)F)F)F)C(=O)O
Quantity 5g
Melting Point 107°C to 110°C
Beilstein 2056612
Formula Weight 226.1
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning