Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4,5,6-Pentafluorophenylacetonitrile, 98%

Click to view available options
Quantity:
1g
5g
653-30-5
C8H2F5N
207.103
MFCD00013814
YDNOJUAQBFXZCR-UHFFFAOYSA-N
2,3,4,5,6-pentafluorophenylacetonitrile,pentafluorophenylacetonitrile,pentafluorophenyl acetonitrile,2-2,3,4,5,6-pentafluorophenyl acetonitrile,2,3,4,5,6-pentafluorobenzyl cyanide,2-perfluorophenyl acetonitrile,benzeneacetonitrile, 2,3,4,5,6-pentafluoro,2-2,3,4,5,6-pentafluorophenyl ethanenitrile,acmc-1ay9x,2-pentafluorophenyl acetonitrile
69554
2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F
653-30-5
C8H2F5N
207.103
MFCD00013814
YDNOJUAQBFXZCR-UHFFFAOYSA-N
2,3,4,5,6-pentafluorophenylacetonitrile,pentafluorophenylacetonitrile,pentafluorophenyl acetonitrile,2-2,3,4,5,6-pentafluorophenyl acetonitrile,2,3,4,5,6-pentafluorobenzyl cyanide,2-perfluorophenyl acetonitrile,benzeneacetonitrile, 2,3,4,5,6-pentafluoro,2-2,3,4,5,6-pentafluorophenyl ethanenitrile,acmc-1ay9x,2-pentafluorophenyl acetonitrile
69554
2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F
Applications
2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Boiling Point 110°C to 111°C (17mmHg)
Quantity 1g
Flash Point >110°C (230°F)
UN Number UN3276
Beilstein 1882341
Formula Weight 207.1
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.