Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
653-34-9,123-31-9
C8H3F5
194.1
MFCD00000300
LVJZCPNIJXVIAT-UHFFFAOYSA-N
2,3,4,5,6-pentafluorostyrene,pentafluorostyrene,benzene, ethenylpentafluoro,1,2,3,4,5-pentafluoro-6-vinyl-benzene,2',3',4',5',6'-pentafluorostyrene,2-2,3,4,5,6-pentafluorophenyl ethyl,vinylpentafluorobenzene,acmc-1avmt,styrene, 2,3,4,5,6-pentafluoro,1,2,3,4,5-pentafluoro-6-vinylbenzene
69556
1-ethenyl-2,3,4,5,6-pentafluorobenzene
C=CC1=C(C(=C(C(=C1F)F)F)F)F
653-34-9,123-31-9
C8H3F5
194.1
MFCD00000300
LVJZCPNIJXVIAT-UHFFFAOYSA-N
2,3,4,5,6-pentafluorostyrene,pentafluorostyrene,benzene, ethenylpentafluoro,1,2,3,4,5-pentafluoro-6-vinyl-benzene,2',3',4',5',6'-pentafluorostyrene,2-2,3,4,5,6-pentafluorophenyl ethyl,vinylpentafluorobenzene,acmc-1avmt,styrene, 2,3,4,5,6-pentafluoro,1,2,3,4,5-pentafluoro-6-vinylbenzene
69556
1-ethenyl-2,3,4,5,6-pentafluorobenzene
C=CC1=C(C(=C(C(=C1F)F)F)F)F
Color Colorless to Yellow
Stabilizer 0.01% tert.-butylcatechol
Specific Gravity 1.406
Packaging Glass bottle
Quantity 5g
Boiling Point 62.0°C to 63.0°C (50.0mmHg)
Infrared Spectrum Authentic
Refractive Index 1.4445 to 1.4465
Flash Point 34°C
Density 1.4060g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C6F5CH=CH2
Formula Weight 194.1
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene, 97%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai

GHS Signal Word: Warning

EINECSNumber : 211-500-5

TSCA : TSCA