Alfa Aesar™ 2,3,4,5,6-Pentamethylbenzyl alcohol, 96%

Manufacturer: Alfa Aesar™ B2257414

Quantity 25g

 View more versions of this product

Catalog No. AAB2257414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 484-66-2
    Molecular Formula C12H18O
    Formula Weight 178.28
    MDL Number MFCD00014421
    Beilstein 2208765
    Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol

    Specifications

    Formula Weight 178.28
    Melting Point 158° to 160°C
    Molecular Formula C12H18O
    Quantity 25g
    MDL Number MFCD00014421
    CAS 484-66-2
    Beilstein 2208765
    Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit