Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,3,4,5,6-Pentamethylbenzyl alcohol, 96%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 1g |
Melting Point | 158°C to 160°C |
Beilstein | 2208765 |
Formula Weight | 178.28 |
Percent Purity | 96% |
Chemical Name or Material | 2,3,4,5,6-Pentamethylbenzyl alcohol |
Safety and Handling
EINECSNumber : 207-609-2
TSCA : No
Recommended Storage : Ambient temperatures