Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentamethylbenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
484-66-2
C12H18O
178.275
MFCD00014421
CMBCAWNOBIGGTE-UHFFFAOYSA-N
2,3,4,5,6-pentamethylbenzyl alcohol,2,3,4,5,6-pentamethylphenyl methanol,pentamethylphenyl methanol,2,3,4,5,6-pentamethylphenyl methan-1-ol,2,3,4,5,6-pentamethylbenzylalcohol,acmc-20aoix,pentamethylbenzyl alcohol,maybridge1_002919,2,3,4,5,6-pentamethyl benzyl alcohol,benzenemethanol,2,3,4,5,6-pentamethyl
1241629
(2,3,4,5,6-pentamethylphenyl)methanol
CC1=C(C(=C(C(=C1C)C)CO)C)C
484-66-2
C12H18O
178.275
MFCD00014421
CMBCAWNOBIGGTE-UHFFFAOYSA-N
2,3,4,5,6-pentamethylbenzyl alcohol,2,3,4,5,6-pentamethylphenyl methanol,pentamethylphenyl methanol,2,3,4,5,6-pentamethylphenyl methan-1-ol,2,3,4,5,6-pentamethylbenzylalcohol,acmc-20aoix,pentamethylbenzyl alcohol,maybridge1_002919,2,3,4,5,6-pentamethyl benzyl alcohol,benzenemethanol,2,3,4,5,6-pentamethyl
1241629
(2,3,4,5,6-pentamethylphenyl)methanol
CC1=C(C(=C(C(=C1C)C)CO)C)C
Quantity 1g
Packaging Glass bottle
Melting Point 159°C to 163°C
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6(CH3)5CH2OH
Formula Weight 178.27
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,3,4,5,6-Pentamethylbenzyl alcohol
Show More Show Less