Alfa Aesar™ 2,3,4,5-Tetrafluorobenzonitrile, 97%

Manufacturer: Alfa Aesar™ B2281314

Quantity 25g

 View more versions of this product

Catalog No. AAB2281314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16582-93-7
    Molecular Formula C7HF4N
    Formula Weight 175.09
    MDL Number MFCD00040187
    UN Number UN3276
    Beilstein 3092489
    Chemical Name or Material 2,3,4,5-Tetrafluorobenzonitrile

    Specifications

    Formula Weight 175.09
    MDL Number MFCD00040187
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Boiling Point 164° to 165°C
    Odor Characteristic
    Quantity 25g
    Molecular Formula C7HF4N
    UN Number UN3276
    Beilstein 3092489
    Health Hazard 1 Danger
    Flash Point 70°C (158°F)
    CAS 16582-93-7
    Health Hazard 2 GHS H Statement
    H311-H227-H302-H332-H315-H319
    Toxic in contact with skin.
    Combustible liquid.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Chemical Name or Material 2,3,4,5-Tetrafluorobenzonitrile
    Assay 97%

    GHS H Statement
    H311-H227-H302-H332-H315-H319
    Toxic in contact with skin.
    Combustible liquid.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit