Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
16582-93-7
C7HF4N
175.086
MFCD00040187
GLTGXIGJLCSEAM-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzenecarbonitrile,tetrafluorobenzonitrile,pubchem9013,intermediates-zcf02243,acmc-1c97m,gltgxigjlcseam-uhfffaoysa,2,3,4,5-tetrafluorobenzonitrile_x000d_,2,3,4,5-tetrakis fluoranyl benzenecarbonitrile
600224
2,3,4,5-tetrafluorobenzonitrile
C1=C(C(=C(C(=C1F)F)F)F)C#N
16582-93-7
C7HF4N
175.086
MFCD00040187
GLTGXIGJLCSEAM-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzenecarbonitrile,tetrafluorobenzonitrile,pubchem9013,intermediates-zcf02243,acmc-1c97m,gltgxigjlcseam-uhfffaoysa,2,3,4,5-tetrafluorobenzonitrile_x000d_,2,3,4,5-tetrakis fluoranyl benzenecarbonitrile
600224
2,3,4,5-tetrafluorobenzonitrile
C1=C(C(=C(C(=C1F)F)F)F)C#N
Infrared Spectrum Authentic
Color Colorless
Refractive Index 1.4545 to 1.4565 (20°C, 589nm)
Packaging Glass Bottle
Density 1.4630g/mL
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula F4C6HCN
Formula Weight 175.08
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzonitrile, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning