Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzotrifluoride, 97%

Click to view available options
Quantity:
5g
25g
100g
654-53-5
C7HF7
218.074
MFCD01862013
ZWVOHERGWLDGFT-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzotrifluoride,1,2,3,4-tetrafluoro-5-trifluoromethyl benzene,benzene, tetrafluoro trifluoromethyl,alpha,alpha,alpha,2,3,4,5-heptafluorotoluene,acmc-209trj,3,4,5,6-tetrafluorobenzotrifluoride,2,3,4,5-tetrafluoro-benzotrifluoride,1,2,3,4-tetrafluoro-5-trifluoromethyl-benzene,2,3,4,5-tetrafluoro-1-trifluoromethyl benzene
2733690
1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
C1=C(C(=C(C(=C1F)F)F)F)C(F)(F)F
654-53-5
C7HF7
218.074
MFCD01862013
ZWVOHERGWLDGFT-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzotrifluoride,1,2,3,4-tetrafluoro-5-trifluoromethyl benzene,benzene, tetrafluoro trifluoromethyl,alpha,alpha,alpha,2,3,4,5-heptafluorotoluene,acmc-209trj,3,4,5,6-tetrafluorobenzotrifluoride,2,3,4,5-tetrafluoro-benzotrifluoride,1,2,3,4-tetrafluoro-5-trifluoromethyl-benzene,2,3,4,5-tetrafluoro-1-trifluoromethyl benzene
2733690
1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
C1=C(C(=C(C(=C1F)F)F)F)C(F)(F)F
Boiling Point 100°C to 102°C
Flash Point 22°C (71°F)
Quantity 25g
UN Number UN1993
Beilstein 1973474
Formula Weight 218.07
Percent Purity 97%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzotrifluoride

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.