Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzoyl chloride, 98%

Manufacturer:  Alfa Aesar™ B2464414

Quantity 25g

 View more versions of this product

Catalog No. AAB2464414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

94695-48-4
212.53
UN3265
2,3,4,5-Tetrafluorobenzoyl chloride
C7HClF4O
MFCD00075164
5266860

Specifications

94695-48-4
212.53
UN3265
65° to 66°C (10mmHg)
25g
98%
C7HClF4O
MFCD00075164
5266860
90°C (194°F)
2,3,4,5-Tetrafluorobenzoyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit