Alfa Aesar™ 2,3,4,5-Tetrafluorobenzoyl chloride, 98%

Manufacturer: Alfa Aesar™ B2464406

Quantity 5g

 View more versions of this product

Catalog No. AAB2464406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94695-48-4
    Molecular Formula C7HClF4O
    Formula Weight 212.53
    MDL Number MFCD00075164
    UN Number UN3265
    Beilstein 5266860
    Chemical Name or Material 2,3,4,5-Tetrafluorobenzoyl chloride

    Specifications

    Formula Weight 212.53
    Molecular Formula C7HClF4O
    MDL Number MFCD00075164
    CAS 94695-48-4
    UN Number UN3265
    Beilstein 5266860
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 65° to 66°C (10mmHg)
    Flash Point 90°C (194°F)
    Quantity 5g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2,3,4,5-Tetrafluorobenzoyl chloride
    Assay 98%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit