Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzyl bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
53001-71-1
C7H3BrF4
242.999
MFCD01631453
WTKVWOHBTGWNEO-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzyl bromide,1-bromomethyl-2,3,4,5-tetrafluorobenzene,2,3,4,5-tetrafluorobenzylbromide,acmc-209l3c,alpha-bromo-2,3,4,5-tetrafluorotoluene,1-bromomethyl-2,3,4,5-tetrafluoro-benzene,5-bromomethyl-1,2,3,4-tetrafluorobenzene,benzene, 1-bromomethyl-2,3,4,5-tetrafluoro,1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
2734024
1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
C1=C(C(=C(C(=C1F)F)F)F)CBr
53001-71-1
C7H3BrF4
242.999
MFCD01631453
WTKVWOHBTGWNEO-UHFFFAOYSA-N
2,3,4,5-tetrafluorobenzyl bromide,1-bromomethyl-2,3,4,5-tetrafluorobenzene,2,3,4,5-tetrafluorobenzylbromide,acmc-209l3c,alpha-bromo-2,3,4,5-tetrafluorotoluene,1-bromomethyl-2,3,4,5-tetrafluoro-benzene,5-bromomethyl-1,2,3,4-tetrafluorobenzene,benzene, 1-bromomethyl-2,3,4,5-tetrafluoro,1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
2734024
1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
C1=C(C(=C(C(=C1F)F)F)F)CBr
Color Colorless to Yellow
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 242.99
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzyl bromide, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA