Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzoyl-D-glucopyranose 90.0+%, TCI America™

Click to view available options
Quantity:
250mg
1g
64768-20-3
C34H28O10
596.588
MFCD00066004
FCDYAJBVISGNLC-UCDCFHRCSA-N
22215583
[(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-hydroxyoxan-2-yl]methyl benzoate
C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
64768-20-3
C34H28O10
596.588
MFCD00066004
FCDYAJBVISGNLC-UCDCFHRCSA-N
22215583
[(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-hydroxyoxan-2-yl]methyl benzoate
C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
Color White-Yellow
Quantity 1g
Melting Point 122°C
Formula Weight 596.59
Physical Form Crystal-Powder at 20°C
Percent Purity ≥90.0% (HPLC)
Chemical Name or Material 2,3,4,6-Tetra-O-benzoyl-D-glucopyranose