Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl p-Trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate

Click to view available options
Quantity:
200mg
1g
468095-63-8
C50H45F6NO6S
901.961
MFCD06797177
LNIPCBRVKJJXQX-ZTQXLAMASA-N
11205261
[(2R,3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-yl] N-[4-(trifluoromethyl)phenyl]-1-[[4-(trifluoromethyl)phenyl]methylsulfanyl]methanimidate
C1=CC=C(C=C1)COCC2C(C(C(C(O2)OC(=NC3=CC=C(C=C3)C(F)(F)F)SCC4=CC=C(C=C4)C(F)(F)F)OCC5=CC=CC=C5)OCC6=CC=CC=C6)OCC7=CC=CC=C7
468095-63-8
C50H45F6NO6S
901.961
MFCD06797177
LNIPCBRVKJJXQX-ZTQXLAMASA-N
11205261
[(2R,3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-yl] N-[4-(trifluoromethyl)phenyl]-1-[[4-(trifluoromethyl)phenyl]methylsulfanyl]methanimidate
C1=CC=C(C=C1)COCC2C(C(C(C(O2)OC(=NC3=CC=C(C=C3)C(F)(F)F)SCC4=CC=C(C=C4)C(F)(F)F)OCC5=CC=CC=C5)OCC6=CC=CC=C6)OCC7=CC=CC=C7
Color White
Quantity 1g
Melting Point 123°C
Formula Weight 901.96
Physical Form Crystal-Powder at 20°C
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl p-Trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate

TSCA : No

Recommended Storage : Freezer