Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%, Thermo Scientific Chemicals

Catalog Number p-7059877
Click to view available options
Quantity:
1 g
5 g
53081-25-7
C34H36O6
MFCD00132927
53081-25-7
C34H36O6
MFCD00132927

2,3,4,6-Tetra-O-benzyl-D-galactopyranose is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 540.65
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-galactopyranose
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit