Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
53081-25-7
C34H36O6
MFCD00132927
53081-25-7
C34H36O6
MFCD00132927
Specific Rotation Condition + 11.00 (20.00°C 589nm,c=1,CHCl3)
Quantity 250mg
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Specific Rotation +9° to +13° (20°C, 589nm) (c=1, CHCl3) after 24 hours
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 540.66
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%
Show More Show Less