Alfa Aesar™ 2-(3,4-Difluorophenyl)-1,3-dioxolane, 98%

Manufacturer: Alfa Aesar™ H3281603

Quantity 1g

 View more versions of this product

Catalog No. AAH3281603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 773101-62-5
    Molecular Formula C9H8F5O2
    Formula Weight 186.16
    MDL Number MFCD06209066
    Synonym 3,4-Difluorobenzaldehyde ethylene ketal
    Chemical Name or Material 2-(3,4-Difluorophenyl)-1,3-dioxolane

    Specifications

    Formula Weight 186.16
    Molecular Formula C9H8F5O2
    MDL Number MFCD06209066
    CAS 773101-62-5
    Synonym 3,4-Difluorobenzaldehyde ethylene ketal
    Quantity 1g
    Chemical Name or Material 2-(3,4-Difluorophenyl)-1,3-dioxolane
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit