Alfa Aesar™ 2-(3,4-Difluorophenyl)ethanol, 97%

Manufacturer: Alfa Aesar™ H3171003

Quantity 1g

 View more versions of this product

Catalog No. AAH3171003

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 286440-92-4
    Molecular Formula C8H8F2O
    Formula Weight 158.15
    MDL Number MFCD00143061
    Synonym 3,4-Difluorophenethyl alcohol
    Chemical Name or Material 2-(3,4-Difluorophenyl)ethanol

    Specifications

    Formula Weight 158.15
    Molecular Formula C8H8F2O
    MDL Number MFCD00143061
    CAS 286440-92-4
    Synonym 3,4-Difluorophenethyl alcohol
    Quantity 1g
    Chemical Name or Material 2-(3,4-Difluorophenyl)ethanol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit