Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(3,4-Dimethoxyphenyl)acetamidoxime, 97%

Click to view available options
Quantity:
1g
5g
42191-48-0
C10H14N2O3
210.233
WJFPRBYVVFXVFB-UHFFFAOYSA-N
2-3,4-dimethoxy-phenyl-n-hydroxy-acetamidine,2-3,4-dimethoxyphenyl-n'-hydroxyethanimidamide,1z-2-3,4-dimethoxyphenyl-n'-hydroxyethanimidamide,benzeneethanimidamide, n-hydroxy-3,4-dimethoxy,benzeneethanimidamide,n-hydroxy-3,4-dimethoxy
16778348
2-(3,4-dimethoxyphenyl)-N'-hydroxyethanimidamide
COC1=C(C=C(C=C1)CC(=NO)N)OC
42191-48-0
C10H14N2O3
210.233
WJFPRBYVVFXVFB-UHFFFAOYSA-N
2-3,4-dimethoxy-phenyl-n-hydroxy-acetamidine,2-3,4-dimethoxyphenyl-n'-hydroxyethanimidamide,1z-2-3,4-dimethoxyphenyl-n'-hydroxyethanimidamide,benzeneethanimidamide, n-hydroxy-3,4-dimethoxy,benzeneethanimidamide,n-hydroxy-3,4-dimethoxy
16778348
2-(3,4-dimethoxyphenyl)-N'-hydroxyethanimidamide
COC1=C(C=C(C=C1)CC(=NO)N)OC
Quantity 5g
Color White
Formula Weight 210.23
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)acetamidoxime