Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[3,4-(Methylenedioxy)phenyl]ethylamine, 95%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
1484-85-1
C9H11NO2
165.192
MFCD00060509
RRIRDPSOCUCGBV-UHFFFAOYSA-N
homopiperonylamine,3,4-methylenedioxyphenethylamine,methylenedioxyphenethylamine,1,3-benzodioxole-5-ethanamine,2-1,3-benzodioxol-5-yl ethanamine,2-benzo d 1,3 dioxol-5-yl ethanamine,methylenedioxyphenylethylamine,benzo-1,3-dioxole-5-ethylamine,3,4-methylenedioxyphenyl ethylamine,phenethylamine, 3,4-methylenedioxy
73874
2-(1,3-benzodioxol-5-yl)ethanamine
C1OC2=C(O1)C=C(C=C2)CCN
1484-85-1
C9H11NO2
165.192
MFCD00060509
RRIRDPSOCUCGBV-UHFFFAOYSA-N
homopiperonylamine,3,4-methylenedioxyphenethylamine,methylenedioxyphenethylamine,1,3-benzodioxole-5-ethanamine,2-1,3-benzodioxol-5-yl ethanamine,2-benzo d 1,3 dioxol-5-yl ethanamine,methylenedioxyphenylethylamine,benzo-1,3-dioxole-5-ethylamine,3,4-methylenedioxyphenyl ethylamine,phenethylamine, 3,4-methylenedioxy
73874
2-(1,3-benzodioxol-5-yl)ethanamine
C1OC2=C(O1)C=C(C=C2)CCN
Sensitivity Air Sensitive
Color Colorless to Yellow
Quantity 5g
Beilstein 383838
Formula Weight 165.19
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-[3,4-(Methylenedioxy)phenyl]ethylamine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SH9670000

storageNote1 : -30° to -10°C