Alfa Aesar™ 2,3,4-Trifluoroaniline, 98%

Manufacturer: Alfa Aesar™ A1025406

Quantity 5g

 View more versions of this product

Catalog No. AAA1025406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3862-73-5
    Molecular Formula C6H4F3N
    Formula Weight 147.1
    MDL Number MFCD00011737
    UN Number UN3082
    Beilstein 3245609
    Chemical Name or Material 2,3,4-Trifluoroaniline

    Specifications

    Formula Weight 147.1
    Health Hazard 2 GHS H Statement
    H318-H373-H302-H312-H227-H315
    Causes serious eye damage.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P280-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 3862-73-5
    Beilstein 3245609
    Flash Point 68°C (154°F)
    Boiling Point 90° to 92°C (48mmHg)
    Quantity 5g
    MDL Number MFCD00011737
    UN Number UN3082
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    RTECS Number CY1211500
    Molecular Formula C6H4F3N
    Chemical Name or Material 2,3,4-Trifluoroaniline
    Assay 98%

    GHS H Statement
    H318-H373-H302-H312-H227-H315
    Causes serious eye damage.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.
    Causes skin irritation.

    GHS P Statement
    P210-P260-P280-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit