Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4-Trifluoroaniline, 98%

Manufacturer:  Alfa Aesar™ A1025414

Quantity 25g

 View more versions of this product

Catalog No. AAA1025414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3862-73-5
147.1
UN3082
2,3,4-Trifluoroaniline
C6H4F3N
MFCD00011737
3245609

Specifications

68°C (154°F)
25g
C6H4F3N
MFCD00011737
3245609
98%
90° to 92°C (48mmHg)
3862-73-5
147.1
UN3082
2,3,4-Trifluoroaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H373-H302-H312-H227-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Causes skin irritation.

GHS P Statement
P210-P260-P280-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

RTECSNumber : CY1211500

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit