Alfa Aesar™ 2,3,4-Trifluoroanisole, 97+%

Manufacturer: Alfa Aesar™ B2401403

Quantity 1g

 View more versions of this product

Catalog No. AAB2401403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 203245-16-3
    Molecular Formula C7H5F3O
    Formula Weight 162.11
    MDL Number MFCD00042396
    Synonym 1-Methoxy-2,3,4-trifluorobenzene
    Chemical Name or Material 2,3,4-Trifluoroanisole

    Specifications

    Formula Weight 162.11
    MDL Number MFCD00042396
    CAS 203245-16-3
    Synonym 1-Methoxy-2,3,4-trifluorobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Flash Point 61°C (142°F)
    Quantity 1g
    Molecular Formula C7H5F3O
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,3,4-Trifluoroanisole
    Assay ≥97%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit