Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trifluorobenzotrifluoride, 98%, Acros Organics

Click to view available options
Quantity:
1g
393-01-1
C7H2F6
200.083
MFCD02683042
KJOHHPPNYVMWRE-UHFFFAOYSA-N
2,3,4-trifluorobenzotrifluoride,1,2,3-trifluoro-4-trifluoromethyl benzene,2,3,4-trifluoro-benzotrifluoride,2,3,4-trifluorobenzotrifluoride_x000d_,2,3,4-trifluoro-1-trifluoromethyl benzene,1,2,3-tris fluoranyl-4-trifluoromethyl benzene
2734025
1,2,3-trifluoro-4-(trifluoromethyl)benzene
C1=CC(=C(C(=C1C(F)(F)F)F)F)F
393-01-1
C7H2F6
200.083
MFCD02683042
KJOHHPPNYVMWRE-UHFFFAOYSA-N
2,3,4-trifluorobenzotrifluoride,1,2,3-trifluoro-4-trifluoromethyl benzene,2,3,4-trifluoro-benzotrifluoride,2,3,4-trifluorobenzotrifluoride_x000d_,2,3,4-trifluoro-1-trifluoromethyl benzene,1,2,3-tris fluoranyl-4-trifluoromethyl benzene
2734025
1,2,3-trifluoro-4-(trifluoromethyl)benzene
C1=CC(=C(C(=C1C(F)(F)F)F)F)F
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Quantity 1g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 200.08
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 3, 4-Trifluorobenzotrifluoride, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger