Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,4-Trifluorobenzyl alcohol, 97%

Click to view available options
Quantity:
1g
5g
144284-24-2
C7H5F3O
162.111
MFCD00083551
LOZFHOHUIYBNCJ-UHFFFAOYSA-N
2,3,4-trifluorophenyl methanol,2,3,4-trifluorobenzyl alcohol,2,3,4-trifluorobenzylalcohol,2,3,4-trifluoro benzenemethanol,2,3,4-trifluoro-phenyl-methanol,2,3,4-trifluorophenyl methan-1-ol,pubchem8282,acmc-209vax,benzenemethanol,2,3,4-trifluoro,2,3,4-tris fluoranyl phenyl methanol
2777027
(2,3,4-trifluorophenyl)methanol
C1=CC(=C(C(=C1CO)F)F)F
144284-24-2
C7H5F3O
162.111
MFCD00083551
LOZFHOHUIYBNCJ-UHFFFAOYSA-N
2,3,4-trifluorophenyl methanol,2,3,4-trifluorobenzyl alcohol,2,3,4-trifluorobenzylalcohol,2,3,4-trifluoro benzenemethanol,2,3,4-trifluoro-phenyl-methanol,2,3,4-trifluorophenyl methan-1-ol,pubchem8282,acmc-209vax,benzenemethanol,2,3,4-trifluoro,2,3,4-tris fluoranyl phenyl methanol
2777027
(2,3,4-trifluorophenyl)methanol
C1=CC(=C(C(=C1CO)F)F)F
Quantity 5g
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 2,3,4-Trifluorobenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning