Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trimethoxybenzoic acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
573-11-5
C10H12O5
212.2
MFCD00002433
HZNQSWJZTWOTKM-UHFFFAOYSA-N
benzoic acid, 2,3,4-trimethoxy,2,3,4-trimethoxybenzoicacid,2,3,4-trimethoxy-benzoic acid,trimethoxybenzoic acid,benzoic acid, trimethoxy,acmc-209lx4,ksc265s3r,2,3,4-trimethoxy benzoic acid,2,3,4-trimethoxybenzoic acid
11308
2,3,4-trimethoxybenzoic acid
COC1=C(C(=C(C=C1)C(=O)O)OC)OC
573-11-5
C10H12O5
212.2
MFCD00002433
HZNQSWJZTWOTKM-UHFFFAOYSA-N
benzoic acid, 2,3,4-trimethoxy,2,3,4-trimethoxybenzoicacid,2,3,4-trimethoxy-benzoic acid,trimethoxybenzoic acid,benzoic acid, trimethoxy,acmc-209lx4,ksc265s3r,2,3,4-trimethoxy benzoic acid,2,3,4-trimethoxybenzoic acid
11308
2,3,4-trimethoxybenzoic acid
COC1=C(C(=C(C=C1)C(=O)O)OC)OC
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 99°C to 102°C
Color White
Quantity 25g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CH3O)3C6H2CO2H
Beilstein 10, 465
Solubility Information Solubility in water: insoluble
Formula Weight 212.2
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 2, 3, 4-Trimethoxybenzoic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 209-350-